Learn More
Gibco™ Human HPRG Recombinant Protein, PeproTech®
Recombinant Protein
87.00€ - 3215.00€
Especificaciones
Número de acceso P04196
Para utilizar con (aplicación) Functional Assay
Formulación protein with no preservative
ID de gen (Entrez) 3273
Peso molecular 57.7 kDa
Ver más especificaciones
Productos 6
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
17871373
Ver documentos
Gibco™
100-60-100UG
100 μg
646.00€
100 microgramos
17820793
Ver documentos
Gibco™
100-60-1MG
1 mg
3215.00€
1 miligramo
17800883
Ver documentos
Gibco™
100-60-250UG
250 μg
1126.00€
250 microgramos
17871463
Ver documentos
Gibco™
100-60-25UG
25 μg
211.00€
25 microgramos
17820313
Ver documentos
Gibco™
100-60-500UG
500 μg
1922.00€
500 microgramos
17891463
Ver documentos
Gibco™
100-60-5UG
5 μg
87.00€
5 microgramos
Descripción
Descripción
Recombinant Human HPRG is a 57.7 kDa glycoprotein containing 507 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
HRG contains two cystatin-like domains and is located in plasma and platelets. The physiological function has not been determined but it is known that the protein binds heme, dyes and divalent metal ions. It can inhibit rosette formation and interacts with heparin, thrombospondin and plasminogen. Two of the protein's effects, the inhibition of fibrinolysis and the reduction of inhibition of coagulation, indicate a potential prothrombotic effect. Mutations in this gene lead to thrombophilia due to abnormal histidine-rich glycoprotein levels.
Especificaciones
Mostrar más
Documentos
Product Certifications
Título del producto
Gibco™ Human HPRG Recombinant Protein, PeproTech®