missing translation for 'onlineSavingsMsg'
Learn More
Gibco™ Human HPRG Recombinant Protein, PeproTech® Código de producto.: 17800883

Gibco™ Human HPRG Recombinant Protein, PeproTech®

Artikelnummer. 17800883
250 μg, 250 microgramos
missing translation for 'orderingAttributeHoverText'
Cantidad:
1 mg
100 μg
25 μg
250 μg
500 μg
5 μg
missing translation for 'unitSize'
1 miligramo
100 microgramos
25 microgramos
250 microgramos
5 microgramos
500 microgramos
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Código de producto. 17800883

Marca: Gibco™ 10060250UG

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Recombinant Protein

Recombinant Human HPRG is a 57.7 kDa glycoprotein containing 507 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

HRG contains two cystatin-like domains and is located in plasma and platelets. The physiological function has not been determined but it is known that the protein binds heme, dyes and divalent metal ions. It can inhibit rosette formation and interacts with heparin, thrombospondin and plasminogen. Two of the protein's effects, the inhibition of fibrinolysis and the reduction of inhibition of coagulation, indicate a potential prothrombotic effect. Mutations in this gene lead to thrombophilia due to abnormal histidine-rich glycoprotein levels.
TRUSTED_SUSTAINABILITY

Spezifikation

Número de acceso P04196
Para utilizar con (aplicación) Functional Assay
Formulación protein with no preservative
ID de gen (Entrez) 3273
Peso molecular 57.7 kDa
Nombre Human HPRG
Cantidad 250 μg
Fuente CHO cells
Estado normativo RUO
Concentración de endotoxinas <1 EU/ μg
Alias de gen AI265597; AW413091; D16jh2; D18020; DKFZp779H1622; histidine rich glycoprotein; histidine-proline-rich glycoprotein; Histidine-rich glycoprotein; histidine-rich glycoprotein 1; HPRG; HRG; HRG1; Hrgp; THPH11
Nombre común HRG
Símbolo de gen HRG
Actividad biológica Measured by its ability to support adhesion of Con-A activated MOLT-4 cells to the Recombinant Human HPRG-coated surface. The ED50 for this effect is 1.5-2.0 ug/ml
Conjugado Unconjugated
Recombinante Recombinant
Secuencia VSPTDCSAVE PEAEKALDLI NKRRRDGYLF QLLRIADAHL DRVENTTVYY LVLDVQESDC SVLSRKYWND CEPPDSRRPS EIVIGQCKVI ATRHSHESQD LRVIDFNCTT SSVSSALANT KDSPVLIDFF EDTERYRKQA NKALEKYKEE NDDFASFRVD RIERVARVRG GEGTGYFVDF SVRNCPRHHF PRHPNVFGFC RADLFYDVEA LDLESPKNLV INCEVFDPQE HENINGVPPH LGHPFHWGGH ERSSTTKPPF KPHGSRDHHH PHKPHEHGPP PPPDERDHSH GPPLPQGPPP LLPMSCSSCQ HATFGTNGAQ RHSHNNNSSD LHPHKHHSHE QHPHGHHPHA HHPHEHDTHR QHPHGHHPHG HHPHGHHPHG HHPHGHHPHC HDFQDYGPCD PPPHNQGHCC HGHGPPPGHL RRRGPGKGPR PFHCRQIGSV YRLPPLRKGE VLPLPEANFP SFPLPHHKHP LKPDNQPFPQ SVSESCPGKF KSGFPQVSMF FTHTFPK
Contenido y almacenamiento -20°C
Sistema de expresión CHO cells
Formulario Lyophilized
Grado de pureza o calidad ≥ 90% by SDS-PAGE gel and HPLC analyses.
Mehr anzeigen Weniger anzeigen
Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts
Gibco™ Human HPRG Recombinant Protein, PeproTech® >

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt