missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human HPRG Recombinant Protein, PeproTech®

Código de producto. 17871373
Cambiar vista
Click to view available options
Cantidad:
100 μg
1 mg
250 μg
25 μg
500 μg
5 μg
Tamaño de la unidad:
1 miligramo
100 microgramos
25 microgramos
250 microgramos
5 microgramos
500 microgramos
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
17871373 100 μg 100 microgramos
17820793 1 mg 1 miligramo
17800883 250 μg 250 microgramos
17871463 25 μg 25 microgramos
17820313 500 μg 500 microgramos
17891463 5 μg 5 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 17871373 Proveedor Gibco™ N.º de proveedor 10060100UG

Please to purchase this item. Need a web account? Register with us today!

Este artículo no se puede devolver. Vea la política de devoluciones

Recombinant Protein

Recombinant Human HPRG is a 57.7 kDa glycoprotein containing 507 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

HRG contains two cystatin-like domains and is located in plasma and platelets. The physiological function has not been determined but it is known that the protein binds heme, dyes and divalent metal ions. It can inhibit rosette formation and interacts with heparin, thrombospondin and plasminogen. Two of the protein's effects, the inhibition of fibrinolysis and the reduction of inhibition of coagulation, indicate a potential prothrombotic effect. Mutations in this gene lead to thrombophilia due to abnormal histidine-rich glycoprotein levels.
TRUSTED_SUSTAINABILITY

Especificaciones

Número de acceso P04196
Para utilizar con (aplicación) Functional Assay
Formulación protein with no preservative
ID de gen (Entrez) 3273
Peso molecular 57.7 kDa
Nombre Human HPRG
Cantidad 100 μg
Fuente CHO cells
Estado normativo RUO
Concentración de endotoxinas <1 EU/ μg
Alias de gen AI265597; AW413091; D16jh2; D18020; DKFZp779H1622; histidine rich glycoprotein; histidine-proline-rich glycoprotein; Histidine-rich glycoprotein; histidine-rich glycoprotein 1; HPRG; HRG; HRG1; Hrgp; THPH11
Nombre común HRG
Símbolo de gen HRG
Actividad biológica Measured by its ability to support adhesion of Con-A activated MOLT-4 cells to the Recombinant Human HPRG-coated surface. The ED50 for this effect is 1.5-2.0 ug/ml
Conjugado Unconjugated
Recombinante Recombinant
Secuencia VSPTDCSAVE PEAEKALDLI NKRRRDGYLF QLLRIADAHL DRVENTTVYY LVLDVQESDC SVLSRKYWND CEPPDSRRPS EIVIGQCKVI ATRHSHESQD LRVIDFNCTT SSVSSALANT KDSPVLIDFF EDTERYRKQA NKALEKYKEE NDDFASFRVD RIERVARVRG GEGTGYFVDF SVRNCPRHHF PRHPNVFGFC RADLFYDVEA LDLESPKNLV INCEVFDPQE HENINGVPPH LGHPFHWGGH ERSSTTKPPF KPHGSRDHHH PHKPHEHGPP PPPDERDHSH GPPLPQGPPP LLPMSCSSCQ HATFGTNGAQ RHSHNNNSSD LHPHKHHSHE QHPHGHHPHA HHPHEHDTHR QHPHGHHPHG HHPHGHHPHG HHPHGHHPHC HDFQDYGPCD PPPHNQGHCC HGHGPPPGHL RRRGPGKGPR PFHCRQIGSV YRLPPLRKGE VLPLPEANFP SFPLPHHKHP LKPDNQPFPQ SVSESCPGKF KSGFPQVSMF FTHTFPK
Contenido y almacenamiento -20°C
Sistema de expresión CHO cells
Formulario Lyophilized
Grado de pureza o calidad ≥ 90% by SDS-PAGE gel and HPLC analyses.
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.