missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human STC-1 / Stanniocalcin-1 Protein
A DNA sequence encoding the Homo sapiens (Human) STC-1 / Stanniocalcin-1, was expressed in the hosts and tags indicated.
160.00€ - 1855.00€
Especificaciones
Número de acceso | NP_003146.1 |
---|---|
ID de gen (Entrez) | 6781 |
Nombre | STC-1 / Stanniocalcin-1 Protein |
Estado normativo | Research Use Only |
Concentración de endotoxinas | Not determined. |
Product Code | Brand | Cantidad | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Cantidad | Price | Quantity & Availability | |||||
16075911
|
enQuireBio™
QP8677-ec-10ug |
10 μg | N/A | ||||||
16015921
|
enQuireBio™
QP8677-ec-50ug |
50 μg | N/A | ||||||
16065911
|
enQuireBio™
QP8677-ec-100ug |
100 μg | N/A | ||||||
16095911
|
enQuireBio™
QP8677-ec-200ug |
200 μg | N/A | ||||||
16005921
|
enQuireBio™
QP8677-ec-500ug |
500 μg | N/A | ||||||
16085911
|
enQuireBio™
QP8677-ec-1mg |
1 mg | N/A | ||||||
Specifications
NP_003146.1 | |
STC-1 / Stanniocalcin-1 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
6781 | |
Research Use Only | |
STC1 | |
Recombinant Protein | |
E. coli | |
SAEVVRCLNS ALQVGCGAFA CLENSTCDTD GMYDICKSFL YSAAKFDTQG KAFVKESLKC IANGVTSKVF LAIRRCSTFQ RMIAEVQEEC YSKLNVCSIA KRNPEAITEV VQLPNHFSNR YYNRLVRSLL ECDEDTVSTI RDSLMEKIGP NMASLFHILQ TDHCAQTHPR ADFNRRRTNE PQKLKVLLRN LRGEEDSPSH IKRTSHESA | |
Greater than 80% as determined by SDS-PAGE. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Human STC-1 / Stanniocalcin-1 Protein