Learn More
Micronova™ Toallita en manopla para entorno controlado esponja fina doble de 305 mm NovaPoly PolyMitt
MittWipes™ Wipers offer a high-quality wiper with the security of a glove. The basic mitt fits over a gloved hand, allowing easier cleaning of hard to reach areas around carts, shelving, tanks and tubing. Can also be used as a protective layer over standard gloves.
Marca: Micronova™ MWDT-611
Descripción
- Microfibre
- Double thin foam interior for increased absorbency
- Fits comfortably over a gloved hand
- Can be used as a protective layer over standard gloves in some clean build and maintenance applications
Especificaciones
Poliéster | |
Blanco | |
Fits comfortably over a gloved hand, for easy cleaning of hard-to-reach areas around carts, shelving, tanks and tubing. Can also be used as a protective layer over standard gloves | |
Foam interior |
Certificados
Se requiere un número de lote para mostrar resultados para certificados. Para encontrar su número de lote en pedidos anteriores, utilice el área de estado de pedidos.
Número de lote | Tipo de certificado | Fecha | Catalog Number |
---|---|---|---|
049901-000 | Certificado de conformidad | 24/06/2025 | MWDT-611 |
049592-000 | Certificado de conformidad | 16/06/2025 | MWDT-611 |
048972-000 | Certificado de conformidad | 09/04/2025 | MWDT-611 |
048070-000 | Certificado de conformidad | 17/03/2025 | MWDT-611 |
049901-000 | Certificado de calidad | 24/06/2025 | MWDT-611 |