Learn More
Amitriptyline hydrochloride, MedChemExpress
MedChemExpress Amitriptyline hydrochloride is an inhibitor of serotonin reuptake transporter (SERT) and noradrenaline reuptake transporter (NET), with Kis of 3.45 nM and 13.3 nM for human SERT and NET, respectively.

en la oferta - 48.60€
Especificaciones
Nombre del producto químico o material | Amitriptyline hydrochloride |
---|---|
Forma física | Solid |
CAS | 549-18-8 |
Color | Blanco |
Notas de grado de pureza | Research |
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
16450978
![]() |
Medchem Express
HY-B0527A |
5 g |
48.60€
5 g |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | |||||
16432219
![]() |
Medchem Express
HY-B0527A |
1 g |
en la oferta
1 g |
N/A | Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
Description
Amitriptyline hydrochloride also weakly binds to dopamine reuptake transporter (DAT) with a Ki of 2.58 μM. Amitriptyline hydrochloride also inhibits adrenergic, muscarinic, histamine and 5-HT receptors. Amitriptyline hydrochloride is a TrkA and TrkB receptors agonist with potent neurotrophic activity. Amitriptyline hydrochloride has antidepressant activity.Specifications
Amitriptyline hydrochloride | |
549-18-8 | |
Research | |
Neuroscience-Neurodegeneration | |
CN(C)CC/C=C1C2=CC=CC=C2CCC3=C\1C=CC=C3.Cl | |
313.86 | |
Research |
Solid | |
Blanco | |
C20H24ClN | |
DMSO : ≥ 100 mg/mL (318.61 mM) ∣H2O : ≥ 50 mg/mL (159.31 mM) | |
313.86 | |
99.56% |
Safety and Handling
H301∣H319∣H361∣H400
missing translation for 'shelfLife' : 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
Recommended Storage : 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.