missing translation for 'onlineSavingsMsg'
Learn More

Amitriptyline hydrochloride, MedChemExpress

Código de producto. 16432219
Encompass_Preferred
Change view
Click to view available options
Cantidad:
1 g
5 g
Unit Size:
1 g
5 g
Product Code. Cantidad unitSize
16432219 1 g 1 g
16450978 5 g 5 g
2 options
This item is not returnable. View return policy

Product Code. 16432219

Brand: Medchem Express HYB0527A

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo requiere una actualización de precios antes de ser solicitado.

Solicite el precio de este artículo aquí.

This item is not returnable. View return policy

MedChemExpress Amitriptyline hydrochloride is an inhibitor of serotonin reuptake transporter (SERT) and noradrenaline reuptake transporter (NET), with Kis of 3.45 nM and 13.3 nM for human SERT and NET, respectively.

Amitriptyline hydrochloride also weakly binds to dopamine reuptake transporter (DAT) with a Ki of 2.58 μM. Amitriptyline hydrochloride also inhibits adrenergic, muscarinic, histamine and 5-HT receptors. Amitriptyline hydrochloride is a TrkA and TrkB receptors agonist with potent neurotrophic activity. Amitriptyline hydrochloride has antidepressant activity.

Specifications

Nombre del producto químico o material Amitriptyline hydrochloride
Forma física Solid
CAS 549-18-8
Color Blanco
Cantidad 1 g
Notas de grado de pureza Research
Fórmula molecular C20H24ClN
Para utilizar con (aplicación) Neuroscience-Neurodegeneration
Información de solubilidad DMSO : ≥ 100 mg/mL (318.61 mM) ∣H2O : ≥ 50 mg/mL (159.31 mM)
SMILES CN(C)CC/C=C1C2=CC=CC=C2CCC3=C\1C=CC=C3.Cl
Peso molecular (g/mol) 313.86
Formula Weight (peso de la fórmula) 313.86
Porcentaje de pureza 99.56%
Grado Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.