missing translation for 'onlineSavingsMsg'
Learn More

Tribencilamina, +99 %, Thermo Scientific Chemicals

Catalog Number p-7027217
Click to view available options
Cantidad:
25 g
100 g
Este artículo no se puede devolver. Vea la política de devoluciones
620-40-6
C21H21N
287.41
MFCD00004773
MXHTZQSKTCCMFG-UHFFFAOYSA-N
tribenzylamine, benzenemethanamine, n,n-bis phenylmethyl, unii-hz10o1931j, trisbenzylamine, n,n-dibenzyl-1-phenyl-methanamine, pubchem21076, acmc-1atok, dsstox_cid_27031, dsstox_rid_82052
24321
N,N-dibencil-1-fenilmetanamina
C(N(CC1=CC=CC=C1)CC1=CC=CC=C1)C1=CC=CC=C1
Este artículo no se puede devolver. Vea la política de devoluciones
620-40-6
C21H21N
287.41
MFCD00004773
MXHTZQSKTCCMFG-UHFFFAOYSA-N
tribenzylamine, benzenemethanamine, n,n-bis phenylmethyl, unii-hz10o1931j, trisbenzylamine, n,n-dibenzyl-1-phenyl-methanamine, pubchem21076, acmc-1atok, dsstox_cid_27031, dsstox_rid_82052
24321
N,N-dibencil-1-fenilmetanamina
C(N(CC1=CC=CC=C1)CC1=CC=CC=C1)C1=CC=CC=C1

Has been used as a base in peptide synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material Tribenzylamine
Punto de fusión 91°C to 94°C
Punto de ebullición 230°C (13 mmHg)
Temperatura de inflamación 203°C (397°F)
Olor Weak
Cantidad 25 g
Beilstein 2214682
Formula Weight (peso de la fórmula) 287.41
Porcentaje de pureza ≥99%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'