missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TLR3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 17766-1-AP-150UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
TLR3 is involved in Herpes simplex encephalitis 2 (HSE2), a rare complication of human herpesvirus 1 (HHV-1) infection, occurring in only a small minority of HHV-1 infected individuals. HSE is characterized by hemorrhagic necrosis of parts of the temporal and frontal lobes. Onset is over several days and involves fever, headache, seizures, stupor, and often coma, frequently with a fatal outcome. Disease susceptibility is associated with variations affecting the gene represented in this entry. TLR3 mutations predispose otherwise healthy individuals to isolated herpes simplex encephalitis through a mechanism that involves impaired IFNs production and reduced immune defense against viral infection in the central nervous system.Especificaciones
TLR3 | |
Polyclonal | |
Unconjugated | |
TLR3 | |
CD283, TLR3, toll like receptor 3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
142980, 364594, 7098 | |
-20°C | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin) | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O15455, Q99MB1 | |
Tlr3 | |
TLR3 Fusion Protein Ag12171 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Sugerencias de productos
Customers who viewed this item also viewed.
Viewing 1-4 of
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
TLR3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido