Learn More
ACROBiosystems SARS-CoV-2 (COVID-19) Nucleocapsid protein, His Tag

418.20€ - 3034.50€
Especificaciones
Número de acceso | QHO62115.1 |
---|---|
Peso molecular | 47.3 kDa |
Fuente | SARS-CoV-2 Nucleocapsid protein, His Tag (NUN-C5227) is expressed and secreted by human 293 cells (HEK293). It contains AA Met 1 - Ala 419 (Accession # QHO62115.1). |
Inmunógeno | Met 1 - Ala 419 |
Requisitos de almacenamiento | -20°C |
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
16575174
![]() |
ACROBiosystems
NUN-C5227-100UG |
100 μg |
418.20€
100 microgramos |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | |||||
16585174
![]() |
ACROBiosystems
NUN-C5227-1MG |
1 mg |
3034.50€
1 miligramo |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | |||||
Descripción
Nucleocapsid (N) protein is the most abundant protein found in coronavirus. CoV N protein is a highly immunogenic phosphoprotein important for viral genome replication and modulation of cell signaling pathways. It was first identified by a research team while they were screening for ADP-ribosylated proteins during coronavirus (CoV) infection (Grunewald M. E., et al. 2017, Virology; 517: 62-68). The array of diverse functional activities accommodated in N protein makes it more than a structural protein but also an interesting target in the development of antiviral therapeutics. Because of the conservation of N protein sequence and its strong immunogenicity, N protein of coronavirus is chosen as a diagnostic tool.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 47.3 kDa. The protein migrates as 60-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Especificaciones
QHO62115.1 | |
SARS-CoV-2 Nucleocapsid protein, His Tag (NUN-C5227) is expressed and secreted by human 293 cells (HEK293). It contains AA Met 1 - Ala 419 (Accession # QHO62115.1). | |
-20°C | |
Nucleocapsid protein | |
SARS-CoV-2 | |
PBS, pH7.4 | |
HEK293 | |
Viral Proteins |
47.3 kDa | |
Met 1 - Ala 419 | |
Nucleocapsid protein,NP,Protein N,COVID-19 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
El Programa Encompass de Fisher Scientific ofrece artículos que no forman parte de nuestra cartera de distribución. Estos productos no suelen tener imágenes ni descripciones detalladas. Sin embargo, nos comprometemos a mejorar su experiencia de compra. Utilice el siguiente formulario para hacernos llegar sus comentarios sobre el contenido de este producto.