missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Ovine Leptin tA Protein
A cDNA sequence encoding the Leptin tA was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP10767-20ug
Este artículo no se puede devolver.
Vea la política de devoluciones
Especificaciones
Leptin tA Protein | |
Research Use Only | |
Pegylated triple antagonist is capable of inhibiting leptin-induced proliferation of BAF/3 cells stably transfected with the long form of human leptin receptor. Its in vitro activity is 6-8 fold lower than the non-pegylated antagonist but in vivo it has profound weight gain effect (as compared to the non-pegylated antagonist like in mouse leptin antagonists), resulting mainly from increased food intake. | |
Ovine | |
Untagged | |
Greater than 95.0% as determined by: (a) Gel filtration analysis. (b) Analysis by SDS-PAGE. |
20 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
The protein was lyophilized from a concentrated (0.65 mg/ml) solution with 0.003mM NaHCO3. |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
enQuireBio™ Recombinant Ovine Leptin tA Protein > 20μg
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido