Learn More
enQuireBio™ Recombinant Ovine Leptin tA Protein
Código de producto. p-7148576
Click to view available options
Cantidad:
1 mg
20 μg
5 μg
Tamaño de la unidad:
1 miligramo
20 microgramos
5 microgramos
Código de producto. 15915108
Marca: enQuireBio™ QP107675ug
A cDNA sequence encoding the Leptin tA was constructed and used to recombinantly synthesize the protein.
Especificaciones
Nombre Leptin tA Protein
Cantidad 5 μg
Estado normativo Research Use Only
Concentración de endotoxinas < 1.0 EU per ug protein as determined by the LAL method.
Actividad biológica Pegylated triple antagonist is capable of inhibiting leptin-induced proliferation of BAF/3 cells stably transfected with the long form of human leptin receptor. Its in vitro activity is 6-8 fold lower than the non-pegylated antagonist but in vivo it has profound weight gain effect (as compared to the non-pegylated antagonist like in mouse leptin antagonists), resulting mainly from increased food intake.
Tipo de producto Recombinant Protein
Reactividad cruzada Ovine
Especie E. coli
Etiqueta de proteína Untagged
Tampón The protein was lyophilized from a concentrated (0.65 mg/ml) solution with 0.003mM NaHCO3.
Grado de pureza o calidad Greater than 95.0% as determined by: (a) Gel filtration analysis. (b) Analysis by SDS-PAGE.