Learn More
enQuireBio™ Recombinant Mouse Interferon regulatory factor 8 Protein
A DNA sequence encoding the Mus musculus (Mouse) Interferon regulatory factor 8, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6235-ec-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001288740.1 | |
Interferon regulatory factor 8 Protein | |
1 mg | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
15900 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
Irf8 | |
Recombinant Protein | |
E. coli | |
MCDRNGGRRL RQWLIEQIDS SMYPGLIWEN DEKTMFRIPW KHAGKQDYNQ EVDASIFKAW AVFKGKFKEG DKAEPATWKT RLRCALNKSP DFEEVTDRSQ LDISEPYKVY RIVPEEEQKC KLGVAPAGCM SEVPEMECGR SEIEELIKEP SVDEYMGMTK RSPSPPEACR SQILPDWWVQ QPSAGLPLVT GYAAYDTHHS AFSQMVISFY YGGKLVGQAT TTCLEGCRLS LSQPGLPKLY GPDGLEPVCF PTADTIPSER QRQVTRKLFG HLERGVLLHS NRKGVFVKRL CQGRVFCSGN AVVCKGRPNK LERDEVVQVF DTNQFIRELQ QFYATQSRLP DSRVVLCFGE EFPDTVPLRS KLILVQVEQL YARQLVEEAG KSCGAGSLMP ALEEPQPDQA FRMFPDICTS HQRPFFRENQ QITV |