Learn More
enQuireBio™ Recombinant Mouse DEFB1 / Beta-defensin 1 Protein
A DNA sequence encoding the Mus musculus (Mouse) DEFB1 / Beta-defensin 1, was expressed in the hosts and tags indicated.
309.00€ - 3545.00€
Especificaciones
Número de acceso NP_031869.1
ID de gen (Entrez) 13214
Nombre DEFB1 / Beta-defensin 1 Protein
Estado normativo Research Use Only
Concentración de endotoxinas Not determined.
Ver más especificaciones
Products 6
Product Code Brand Cantidad Price Quantity & Availability  
Product Code Brand Cantidad Price Quantity & Availability  
15908729
missing translation for 'viewDocuments'
enQuireBio™
QP5926-ec-10ug
10 μg Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
N/A
15948729
missing translation for 'viewDocuments'
enQuireBio™
QP5926-ec-50ug
50 μg Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
N/A
15998719
missing translation for 'viewDocuments'
enQuireBio™
QP5926-ec-100ug
100 μg
1025.00€
100 microgramos
15928729
missing translation for 'viewDocuments'
enQuireBio™
QP5926-ec-200ug
200 μg Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
N/A
15938729
missing translation for 'viewDocuments'
enQuireBio™
QP5926-ec-500ug
500 μg Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
N/A
15918729
missing translation for 'viewDocuments'
enQuireBio™
QP5926-ec-1mg
1 mg
3545.00€
1 miligramo
Description
Description
Specifications
Specifications
NP_031869.1
DEFB1 / Beta-defensin 1 Protein
Not determined.
Full Length
Mouse
His-SUMO
Tris-based buffer, 50% glycerol
13214
Research Use Only
Defb1
Recombinant Protein
E. coli
DQYKCLQHGG FCLRSSCPSN TKLQGTCKPD KPNCCKS
Greater than 90% as determined by SDS-PAGE.
Videos
SDS
Documents
Documents
Certificates
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Mouse DEFB1 / Beta-defensin 1 Protein
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.