missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse CXCL2 / MIP-2 Protein
Recombinant Protein
Marca: enQuireBio™ QP5489-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_033166.1 | |
CXCL2 / MIP-2 Protein | |
1 mg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
AVVASELRCQ CLKTLPRVDF KNIQSLSVTP PGPHCAQTEV IATLKGGQKV CLDPEAPLVQ KIIQKILNKG KAN |
20310 | |
Greater than 95% as determined by SDS-PAGE. | |
Research Use Only | |
CXCL2 | |
Mouse | |
Untagged | |
10 mM sodium phosphate, pH 7.5 |