missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Mouse CCL21/6Ckine Protein
Tienda Bio Techne Productos
Click to view available options
Cantidad:
100 μg
1 mg
20 μg
500 μg
5 μg
Tamaño de la unidad:
1 miligramo
100 microgramos
20 microgramos
5 microgramos
500 microgramos
Descripción
CCL21 Protein is fully biologically active when compared to standard. The biological activity determined by a chemotaxis bioassay using murine T-lymphocytes is in a concentration range of 10-100 ng/mL.

Especificaciones
Especificaciones
Para utilizar con (aplicación) | PAGE, Bioactivity |
Formulación | Lyophilized from a 0.2 um filtered concentrated solution in 20 mM PB, pH 7.4, 150 mM NaCl. |
ID de gen (Entrez) | 6366 |
Peso molecular | M.W. (Theoretical): 12.1 kDa |
Cantidad | 100 μg |
Requisitos de almacenamiento | Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. |
Concentración de endotoxinas | Less than 1 EU/μg of CCL21/6Ckine as determined by LAL method. |
Alias de gen | 6CkineSmall-inducible cytokine A21, Beta-chemokine exodus-2, chemokine (C-C motif) ligand 21, CKb9, Exodus-2/CCL21, member 21, SCYA21MGC34555, secondary lymphoid tissue chemokine, SLCSecondary lymphoid-tissue chemokine |
Símbolo de gen | CCL21 |
Reconstitución | Recommended to centrifuge prior to opening. Reconstitute in sterile distilled water or aqueous buffer containing 0.1% BSA to a concentration of 0.1-1.0mg/mL. Apportion stock solutions into working aliquots and store at <-20°C |
Mostrar más |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Novus Biologicals™ Recombinant Mouse CCL21/6Ckine Protein >
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido