missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human TrkC Fc Chimera Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Brand: R&D Systems™ 373-TC-050/CF
This item is not returnable.
View return policy
Description
The Recombinant Human TrkC Fc Chimera Protein is derived from NS0. The Recombinant Human TrkC Fc Chimera Protein has been validated for the following applications: Bioactivity.
Specifications
Lyophilized from a 0.2μm filtered solution in PBS. | |
72 kDa (monomer) | |
50 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
<0.1 EU per 1μg of the protein by the LAL method. | |
Recombinant |
4916 | |
TrkC Fc Chimera | |
Mouse myeloma cell line,NS0-derived human TrkC protein, (N-terminus) Human TrkC (Cys32-Asp428) Accession &Num; Q96CY4 DIEGRMD Human IgG1 (Pro100-Lys330) 6-His tag (C-terminus) | |
Disulfide-linked homodimer | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
R&D Systems™ Recombinant Human TrkC Fc Chimera Protein > 50ug; Carrier Free
Spot an opportunity for improvement?Share a Content Correction