missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human CXCL11/I-TAC Protein
Recombinant Protein
Marca: Novus Biologicals™ NBP2-34922-100ug
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
CXCL11 Protein is fully biologically active when compared to standard. The biological activity determined by a chemotaxis bioassay using human IL-2 activated human T-lymphocytes is in a concentration range of 0.1-10 ng/mL.
Especificaciones
O14625, O14625, O14625 | |
PAGE, Bioactivity | |
6373 | |
100 μg | |
Less than 1 EU/ug of CXCL11/I-TAC as determined by LAL method. | |
CXCL11 | |
Biologically Active Proteins, Chemokines and Cytokines, Cytokine Research | |
CXCL11/I-TAC |
LYOPH | |
Lyophilized from a 0.2 um filtered concentrated solution in 20 mM PB, pH 7.4, 100 mM NaCl. | |
8.3 kDa | |
Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. | |
Beta-R1, b-R1, chemokine (C-X-C motif) ligand 11, H174IP9, Interferon gamma-inducible protein 9, Interferon-inducible T-cell alpha chemoattractant, IP-9member 11, ITAC, I-TAC/CXCL11, I-TACMGC102770, small inducible cytokine subfamily B (Cys-X-Cys), member 9B, Small-inducible cytokine B11 | |
Recommended to centrifuge prior to opening. Reconstitute in sterile distilled water or aqueous buffer containing 0.1% BSA to a concentration of 0.1-1.0mg/mL. Apportion stock solutions into working aliquots and store at <-20°C. | |
>97% pure by SDS-PAGE and HPLC |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Novus Biologicals™ Recombinant Human CXCL11/I-TAC Protein > 100 μg
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido