missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human coiled-coil domain containing 53 GST (N-Term) Protein
Tienda Bio Techne Productos
Click to view available options
Cantidad:
10 μg
25 μg
Tamaño de la unidad:
10 microgramos
25 microgramos
Descripción
This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.

Especificaciones
Especificaciones
Para utilizar con (aplicación) | Western Blot, ELISA, PAGE, Protein Array, Immunoaffinity Purification |
Formulación | 50 mM Tris-HCl, 10 mM reduced Glutathione, pH 8.0 in the elution buffer. |
ID de gen (Entrez) | 51019 |
Peso molecular | 47.6 kDa |
Cantidad | 25 μg |
Requisitos de almacenamiento | Store at -80°C. Avoid freeze-thaw cycles. |
Alias de gen | CGI-116, coiled-coil domain containing 53, Coiled-coil domain-containing protein 53, WASH complex subunit CCDC53 |
Símbolo de gen | CCDC53 |
Grado de pureza o calidad | >80% by SDS-PAGE and Coomassie blue staining |
Expresión | coiled-coil domain containing 53 |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Novus Biologicals™ Recombinant Human coiled-coil domain containing 53 GST (N-Term) Protein >
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido