Learn More
enQuireBio™ Recombinant Human CLDN4 Protein
A cDNA sequence encoding the CLDN4 was constructed and used to recombinantly synthesize the protein.
143.00€ - 8190.00€
Especificaciones
Nombre CLDN4 Protein
Estado normativo Research Use Only
Concentración de endotoxinas Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Tipo de producto Recombinant Protein
Reactividad cruzada Human
Ver más especificaciones
Productos 3
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
15917278
Ver documentos
enQuireBio™
QP11441-2ug
2 μg
143.00€
2 microgramos
15997268
Ver documentos
enQuireBio™
QP11441-10ug
10 μg
222.00€
10 microgramos
15907278
Ver documentos
enQuireBio™
QP11441-1mg
1 mg
8190.00€
1 miligramo
Especificaciones
CLDN4 Protein
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
Filtered (0.4 microm) and lyophilized from 0.5 mg/ml in 0.05M Acetate buffer pH 4.
Research Use Only
Recombinant Protein
E. coli
MKHHHHHHAS MWRVTAFIGS NIVTSQTIWE GLWMNCVVQS TGQMQCKVYD SLLALPQDLQ AAR LEVLFQG PTAHNIIQDF YNPLVASGQK REM MWRVTAFIGS NIVTSQTIWE GLWMNCVVQS TGQMQCKVYD SLLALPQDLQ AAR LEVLFQG PTAHNIIQDF YNPLVASGQK REM
Greater than 90.0% as determined by SDS-PAGE.
Documentos
Product Certifications
Título del producto
enQuireBio™ Recombinant Human CLDN4 Protein