Learn More
R&D Systems™ Recombinant Human CCL4/MIP-1 beta Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Marca:  R&D Systems™ 271-BME-050
 Ver más versiones de este producto
Código de producto. 18108700
  • 506.00€ / 50 microgramos
Descripción
The Recombinant Human CCL4/MIP-1 beta Protein is derived from E. coli. The Recombinant Human CCL4/MIP-1 beta Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY
Especificaciones
Especificaciones
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
7.8 kDa & 7.3 kDa
50 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Recombinant
6351
CCL4/MIP-1 beta
E. coli-derived human CCL4/MIP-1 beta protein Ala24-Asn92 (major); Pro30-Asn92 (Minor)
Unconjugated
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Documentos
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
R&D Systems™ Recombinant Human CCL4/MIP-1 beta Protein > 50ug
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.