missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human CCL11/Eotaxin Protein
Recombinant Protein
Marca: Novus Biologicals™ NBP2-34976-1mg
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Eotaxin protein is fully biologically active when compared to standard. The biological activity determined by a chemotaxis bioassay using human peripheral blood eosinophils is in a concentration range of 0.1-10.0 ng/mL.
Especificaciones
P51671, P51671, P51671 | |
PAGE, Bioactivity | |
6356 | |
1 mg | |
Less than 1 EU/ug of CCL11/Eotaxin as determined by LAL method. | |
CCL11 | |
Biologically Active Proteins, Chemokines and Cytokines | |
CCL11/Eotaxin |
LYOPH | |
Lyophilized from a 0.2 um filtered concentrated solution in 20 mM PB, pH 7.4, 150 mM NaCl. | |
8.4 kDa | |
Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. | |
C-C motif chemokine 11, CCL11, chemokine (C-C motif) ligand 11, Eosinophil chemotactic protein, eotaxin, Eotaxin/CCL11, eotaxin-1, MGC22554, SCYA11Small-inducible cytokine A11, small inducible cytokine subfamily A (Cys-Cys), member 11 (eotaxin) | |
Recommended to centrifuge prior to opening. Reconstitute in sterile distilled water or aqueous buffer containing 0.1% BSA to a concentration of 0.1-1.0mg/mL. Apportion stock solutions into working aliquots and store at <-20°C. | |
>97% pure by SDS-PAGE and HPLC |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Novus Biologicals™ Recombinant Human CCL11/Eotaxin Protein > 1 mg
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido