Learn More
enQuireBio™ Recombinant Human C1QTNF6 Protein
A cDNA sequence encoding the C1QTNF6 was constructed and used to recombinantly synthesize the protein.
143.00€ - 8190.00€
Especificaciones
Nombre C1QTNF6 Protein
Estado normativo Research Use Only
Concentración de endotoxinas Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Tipo de producto Recombinant Protein
Reactividad cruzada Human
Ver más especificaciones
Productos 3
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
15916568
Ver documentos
enQuireBio™
QP11233-2ug
2 μg
143.00€
2 microgramos
15996558
Ver documentos
enQuireBio™
QP11233-10ug
10 μg
222.00€
10 microgramos
15906568
Ver documentos
enQuireBio™
QP11233-1mg
1 mg
8190.00€
1 miligramo
Especificaciones
C1QTNF6 Protein
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
0.23 mg/ml in 30mM Acetate Buffer pH 4.
Research Use Only
Recombinant Protein
E. coli
MKHHHHHHAS TFDRAVASGC QRCCDSEDPL DPAHVSSASS SGRPHALPEI RPYINITILK GDKGDPGPMG LPGYMGREGP QGEPGPQGSK GDKGEMGSPG APCQKRFFAF SVGRKTALHS GEDFQTLLFE RVFVNLDGCF DMATGQFAAP LRGIYFFSLN VHSWNYKETY VHIMHNQKEA VILYAQPSER SIMQSQSVML DLAYGDRVWV RLFKRQRENA IYSNDFDTYI TFSGHLIKAEDD
Greater than 95% as determined by SDS PAGE.
Documentos
Product Certifications
Título del producto
enQuireBio™ Recombinant Human C1QTNF6 Protein