missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human beta-NGF / Beta-NGF Protein
Recombinant Protein
Marca: enQuireBio™ QP5351-20ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_002497.2 | |
beta-NGF / Beta-NGF Protein | |
Research Use Only | |
NGF | |
Recombinant Protein | |
E. coli | |
MSSSHPIFHR GEFSVCDSVS VWVGDKTTAT DIKGKEVMVL GEVNINNSVF KQYFFETKCR DPNPVDSGCR GIDSKHWNSY CTTTHTFVKA LTMDGKQAAW RFIRIDTACV CVLSRKAVRR A | |
Greater than 95% as determined by SDS-PAGE. |
4803 | |
20 μg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Shows bioactivity at a concentration of 5 ng/mL in the TF-1 cell proliferation assay. The bioactivity acceptance criteria is set at 2 x 10^5 units/mg. | |
Human | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |