Learn More
Novus Biologicals™ Recombinant Human alpha-Synuclein Aggregate Protein (Biologically Active) Código de producto.: 18681109
Novus Biologicals™ Recombinant Human alpha-Synuclein Aggregate Protein (Biologically Active)
Código de producto. 18681109 Tienda Bio Techne Productos
200 μg, 200 microgramos
Click to view available options
Cantidad:
100 μg
200 μg
500 μg
Tamaño de la unidad:
1 set
200 microgramos
500 microgramos
Código de producto. 18681109
Marca: Novus Biologicals™ NBP254789200ug
Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
Ver productos alternativos
Highly purified and high bioactivity. Generating reliable and reproducible results.
An un-tagged full length Human biologically active Alpha-Synuclein recombinant protein aggregate (pre-formed fibrils, Type 1), NCBI Accession #: NP_000336.1. The Recombinant Human alpha-Synuclein Aggregate Protein is derived from E. coli. The Recombinant Human alpha-Synuclein Aggregate Protein has been validated for the following applications: Western Blot, In vitro assay, In vivo assay, SDS-Page, Bioactivity.
TRUSTED_SUSTAINABILITY
Especificaciones
Para utilizar con (aplicación) In vitro Assay, In vivo Assay, SDS-PAGE, Western Blot
Formulación PBS
ID de gen (Entrez) 6622
Nombre Human alpha-Synuclein Aggregate Protein
Método de purificación >95% pure by SDS-PAGE
Cantidad 200 μg
Fuente E.Coli
Inmunógeno MDVFMKGLSKAKEGVVAAAEKTKQGVAEAAGKTKEGVLYVGSKTKEGVVHGVATVAEKTKEQVTNVGGAVVTGVTAVAQKTVEGAGSIAAATGFVKKDQLGKNEEGAPQEGILEDMPVDPDNEAYEMPSEEGYQDYEPEA
Requisitos de almacenamiento Store at -80°C. Avoid freeze/thaw cycles.
Estado normativo RUO
Alias de gen alpha-Synuclein, Lewy body 4, MGC110988, non A-beta component of AD amyloid, Non-A beta component of AD amyloid, non-A4 component of amyloid, Non-A4 component of amyloid precursor, PARK1, PARK4, synuclein, alpha (non A4 component of amyloid precursor)
Símbolo de gen SNCA
Actividad biológica Endogenous alpha-synuclein phosphorylation. 100uM alpha synuclein protein monomer seeded with 10nM alpha synuclein protein aggregate in 25uM Thioflavin T (PBS pH 7.4, 100uL reaction volume) generated a fluorescence intensity of 13,000 Relative Fluorescence Units after incubation at 37°C with shaking at 600 rpm for 24 hours. Fluorescence was measured by excitation at 450nm and emission at 485nm on a Molecular Devices Gemini XPS microplate reader.
Tipo de producto Recombinant Protein
Conjugado Unconjugated
Reactividad cruzada Human
Recombinante Recombinant
Título del producto
Novus Biologicals™ Recombinant Human alpha-Synuclein Aggregate Protein (Biologically Active) >