Learn More
enQuireBio™ Recombinant HPV HPV 16 Protein
A cDNA sequence encoding the HPV 16 was constructed and used to recombinantly synthesize the protein.
248.00€ - 1815.00€
Especificaciones
Nombre HPV HPV 16 Protein
Estado normativo Research Use Only
Concentración de endotoxinas Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Tipo de producto Recombinant Protein
Reactividad cruzada HPV
Ver más especificaciones
Productos 3
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
15999998
Ver documentos
enQuireBio™
QP12307-100ug
100 μg
248.00€
100 microgramos
15910009
Ver documentos
enQuireBio™
QP12307-500ug
500 μg
943.00€
500 microgramos
15900009
Ver documentos
enQuireBio™
QP12307-1mg
1 mg
1815.00€
1 miligramo
Especificaciones
HPV HPV 16 Protein
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
HPV
GST
Recombinant HPV-16 solution in PBS, 100mM arginine and 3M Urea.
Research Use Only
Recombinant Protein
E. coli
MQVTFIYILVITCYENDVNVYHIFFQMSLWLPSEATVYLPPVPVSKVVSTDEYVARTNIYYHAGTSRLLAVGHPYFPIKKPNNNKILVPKVSGLQYRVFRIHLPDPNKFGFPDTSFYNPDTQRLVWACVGVEVGRGQPLGVGISGHPLLNKLDDTENASAYAANAGVDNRECISMDYKQTQLCLIGCKPPIGEHWGKGSPCTNVAVNPGDCPPLELINTVIQDGDMVHTGFGAMDFTTLQANKSEVPLDICTSICKYPDYIKMVSEPYGDSLFFYLRREQMFVRHLFNRAGTVGENVPDDLYIKGSGSTANLASSNYFPTPSGSMVTSDAQIFNKPYWLQRAQGHNNGICWGNQLFVTVVDTTRSTNMSLCAAISTSETTYKNTNFKEYLRHGEEYDLQFIFQLCKITLTADVMTYIHSMNSTILEDWNFGLQPPPGGTLEDTYRFVTQAIACQKHTPPAPKEDDPLKKYTFWEVNLKEKFSADLDQFPLGRKFLLQAGLKAKPKFTLGKRKATPTTSSTSTTAKRKKRKL
Protein is >90% pure as determined by 10% SDS-PAGE (Coomassie staining).
Documentos
Product Certifications
Título del producto
enQuireBio™ Recombinant HPV HPV 16 Protein