missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant HIV-1 nef, Clade B Protein
A cDNA sequence encoding the HIV-1 nef, Clade B was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12254-EC-100ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
HIV-1 nef, Clade B Protein | |
100 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
HIV | |
Untagged |
Greater than 90.0% as determined by HPLC analysis and SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
Lyophilized with 10% glycerol. |