missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Hepatitis E HEV ORF2 Protein
A cDNA sequence encoding the HEV ORF2 was constructed and used to recombinantly synthesize the protein.
238.00€ - 1815.00€
Especificaciones
Nombre | Hepatitis E HEV ORF2 Protein |
---|---|
Estado normativo | Research Use Only |
Concentración de endotoxinas | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Tipo de producto | Recombinant Protein |
Reactividad cruzada | Hepatitis E |
Especificaciones
Hepatitis E HEV ORF2 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Hepatitis E | |
Beta-galactosidase | |
HEV ORF2 protein is >95% pure as determined by 10% PAGE (coomassie staining). |
Research Use Only | |
Recombinant Protein | |
E. coli | |
25mM Tris-HCl, 1mM EDTA, 0.5M urea and 50% glycerol. |
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
enQuireBio™ Recombinant Hepatitis E HEV ORF2 Protein