missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. Coli TRXR Protein
A cDNA sequence encoding the TRXR was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP10894-25ug
Este artículo no se puede devolver.
Vea la política de devoluciones
Especificaciones
TRXR Protein | |
Research Use Only | |
Specific activity is 4-5 units/ml, and was measured in a coupled assay with DTNB and NADPH. The amount of TNB generated by NADPH was measured in absorbance at 412 nm. | |
E. coli | |
Untagged | |
TRXR E. Coli solution containing 20mM Tris HCl pH 8.0, 1mM DTT, and 10% glycerol. |
25 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MGTTKHSKLL ILGSGPAGYT AAVYAARANL QPVLITGMEK GGQLTTTTEV ENWPGDPNDL TGPLLMERMH EHATKFETEI IFDHINKVDL QNRPFRLNGD NGEYTCDALI IATGASARYL GLPSEEAFKG RGVSACATCD GFFYRNQKVA VIGGGNTAVE EALYLSNIAS EVHLIHRRDG FRAEKILIKR LMDKVENGNI ILHTNRTLEE VTGDQMGVTG VRLRDTQNSD NIESLDVAGL FVAIGHSPNT AIFEGQLELE NGYIKVQSGI HGNATQTSIP GVFAAGDVMD HIYRQAITSA GTGCMAALDA ERYLDGLADA K | |
Greater than 90% as determined by SDS-PAGE. |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
enQuireBio™ Recombinant E. Coli TRXR Protein > 25μg
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido