Learn More
enQuireBio™ Recombinant E. coli Murein tetrapeptide carboxypeptidase Protein
A DNA sequence encoding the Escherichia coli (strain K12) Murein tetrapeptide carboxypeptidase, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6956-50ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_415710.1 | |
Murein tetrapeptide carboxypeptidase Protein | |
50 μg | |
Not determined. | |
Full Length | |
E. coli | |
His | |
Tris-based buffer, 50% glycerol |
945756 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
ldcA | |
Recombinant Protein | |
E. coli | |
MSLFHLIAPS GYCIKQHAAL RGIQRLTDAG HQVNNVEVIA RRCERFAGTE TERLEDLNSL ARLTTPNTIV LAVRGGYGAS RLLADIDWQA LVARQQHDPL LICGHSDFTA IQCGLLAHGN VITFSGPMLV ANFGADELNA FTEHHFWLAL RNETFTIEWQ GEGPTCRAEG TLWGGNLAML ISLIGTPWMP KIENGILVLE DINEHPFRVE RMLLQLYHAG ILPRQKAIIL GSFSGSTPND YDAGYNLESV YAFLRSRLSI PLITGLDFGH EQRTVTLPLG AHAILNNTRE GTQLTISGHP VLKM |