Learn More
enQuireBio™ Recombinant Bacteriophage Attachment protein G3P Protein
A DNA sequence encoding the Enterobacteria phage M13 (Bacteriophage M13) Attachment protein G3P, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP9483-ye-100ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_510891.1 | |
Attachment protein G3P Protein | |
100 μg | |
Not determined. | |
Full Length | |
Bacteriophage | |
His | |
Tris-based buffer, 50% glycerol |
927334 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
III | |
Recombinant Protein | |
Yeast | |
AETVESCLAK PHTENSFTNV WKDDKTLDRY ANYEGCLWNA TGVVVCTGDE TQCYGTWVPI GLAIPENEGG GSEGGGSEGG GSEGGGTKPP EYGDTPIPGY TYINPLDGTY PPGTEQNPAN PNPSLEESQP LNTFMFQNNR FRNRQGALTV YTGTVTQGTD PVKTYYQYTP VSSKAMYDAY WNGKFRDCAF HSGFNEDPFV CEYQGQSSDL PQPPVNAGGG SGGGSGGGSE GGGSEGGGSE GGGSEGGGSG GGSGSGDFDY EKMANANKGA MTENADENAL QSDAKGKLDS VATDYGAAID GFIGDVSGLA NGNGATGDFA GSNSQMAQVG DGDNSPLMNN FRQYLPSLPQ SVECRPFVFS AGKPYEFSID CDKINLFRGV FAFLLYVATF MYVFSTFANI LRNKES |