Learn More
ACROBiosystems Mouse SIRP alpha / CD172a Protein, Fc Tag

316.20€ - 2310.30€
Especificaciones
Número de acceso | BAA13520.1 |
---|---|
Peso molecular | 64.3 kDa |
Fuente | Mouse SIRP alpha, Fc Tag (SIA-M5258) is expressed from human 293 cells (HEK293). It contains AA Lys 32 - Asn 373 (Accession # BAA13520.1). |
Inmunógeno | Lys 32 - Asn 373 |
Requisitos de almacenamiento | -20°C |
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
16438079
![]() |
ACROBiosystems
SIA-M5258-100ug |
100 μg |
316.20€
100 microgramos |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | |||||
16448079
![]() |
ACROBiosystems
SIA-M5258-1mg |
1 mg |
2310.30€
1 miligramo |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | |||||
Description
Tyrosine-protein phosphatase non-receptor type substrate 1 (SHPS1) is also known as CD172 antigen-like family member A (CD172a), Macrophage fusion receptor, MyD-1 antigen, Signal-regulatory protein alpha (SIRPA or SIRP alpha) or p84, is a member of the SIRP family, and also belongs to the immunoglobulin superfamily. SIRP alpha is Ubiquitous and highly expressed in brain. SIRPA / CD172a is immunoglobulin-like cell surface receptor for CD47 and acts as docking protein and induces translocation of PTPN6, PTPN11 and other binding partners from the cytosol to the plasma membrane. SIRPA / SHPS-1 supports adhesion of cerebellar neurons, neurite outgrowth and glial cell attachment and may play a key role in intracellular signaling during synaptogenesis and in synaptic function By similarity. SIRPA / MyD1 involved in the negative regulation of receptor tyrosine kinase-coupled cellular responses induced by cell adhesion, growth factors or insulin and mediates negative regulation of phagocytosis, mast cell activation and dendritic cell activation. CD47 binding prevents maturation of immature dendritic cells and inhibits cytokine production by mature dendritic cells.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 64.3 kDa. The protein migrates as 90-100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Specifications
BAA13520.1 | |
Mouse SIRP alpha, Fc Tag (SIA-M5258) is expressed from human 293 cells (HEK293). It contains AA Lys 32 - Asn 373 (Accession # BAA13520.1). | |
-20°C | |
SIRP alpha | |
Mouse | |
PBS, pH7.4 | |
HEK293 | |
Immune Checkpoint Proteins |
64.3 kDa | |
Lys 32 - Asn 373 | |
SHPS1,SIRPA,CD172A,BIT,MFR,MYD1,P84,PTPNS1 | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.