Learn More
Proteintech™ HumanKine™ Recombinant Human R-Spondin-I Protein
R-Spondin 1 plays several roles in embryonic development including sex phenotype reversal, female sex determination, and ovarian differentiation.
Marca:  Proteintech™ HZ-1328-1000UG
 Ver más versiones de este producto
Código de producto. 17791489
  • 2087.00€ / 1000 microgramos
Descripción
R-Spondin 1 is an approximately 40kDA, glycosylated protein that serves as a major agonist of the Wnt signaling pathway. It regulates the turnover of the LRP6 co-receptor by preventing its internalization by DKK-1. It also promotes stem cell turnover in the mammary glands, intestine, colon, and kidneys. Increased R-Spondin 1 expression has been linked to growth and migration of ovarian cancer cells as well as decreased sensitivity of gliomas to radiation-based therapies (PMID: 17804805, 22439850, 32749219, 30572097).
Especificaciones
Especificaciones
284654
Human
Human
Lyophilized
1000 μg
<1 EU/μg
40 kDa
>95%
Certificados
Promociones
Promociones
Título del producto
Proteintech™ HumanKine™ Recombinant Human R-Spondin-I Protein > Recombinant Human R-Spondin-I Protein; Endotoxin Concentration: <1 EU/μg; Form: Lyophilized; Gene ID (Entrez): 284654; Species: Human; Quantity: 1000 μg