Learn More
Gibco™ Human LAG-1 (CCL4L1) Recombinant Protein, PeproTech®
Recombinant Protein
Marca: Gibco™ 300-58-5UG
78.30 EUR hasta el 2025-06-30
Use el código promocional "24108" para aplicar la oferta.
Descripción
Recombinant Human LAG-1 is a 7.7 kDa protein containing 69 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
LAG-1 is CC chemokine that signals through the CCR5 receptor. LAG-1 is identical to MIP-1beta (ACT II isotype) except for two amino acid substitutions; arginine for histidine at position 22 and serine for glycine at position 47 of the mature protein. LAG-1 chemoattracts monocytes, and exhibits activity as an HIV-suppressive factor.
Especificaciones
Q8NHW4 | |
protein with no preservative | |
7.7 kDa | |
5 μg | |
RUO | |
CCL4L1 | |
Determined by its ability to chemoattract CCR5-transfected cells using a concentration of 0.1-10.0 ng/ml | |
Recombinant | |
-20°C | |
Lyophilized |
Functional Assay | |
388372 | |
Human LAG-1 (CCL4L1) | |
E. coli | |
<1 EU/ μg | |
CCL4L1 | |
Unconjugated | |
APMGSDPPTA CCFSYTARKL PRNFVVDYYE TSSLCSQPAV VFQTKRSKQV CADPSESWVQ EYVYDLELN | |
E. coli | |
≥ 98% by SDS-PAGE gel and HPLC analyses. |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.