Learn More
ACROBiosystems Human IL-5 Protein, His Tag

367.20€ - 2805.00€
Especificaciones
Número de acceso | NP_000870.1 |
---|---|
Peso molecular | 15.0 kDa |
Fuente | Human IL-5, His Tag (IL5-H52H3) is expressed from human 293 cells (HEK293). It contains AA Ile 20 - Ser 134 (Accession # P05113-1). |
Inmunógeno | Ile 20 - Ser 134 |
Requisitos de almacenamiento | -20°C |
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
16417709
![]() |
ACROBiosystems
IL5-H52H3-1mg |
1 mg |
2805.00€
1 miligramo |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | |||||
16427709
![]() |
ACROBiosystems
IL5-H52H3-50ug |
50 μg |
367.20€
50 microgramos |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | |||||
Descripción
Interleukin 5 (IL5) is an interleukin produced by type-2 T helper cells and mast cells. IL-5 is a 115-amino acid (in human, 133 in the mouse) -long TH2 cytokine that is part of the hematopoietic family. Unlike other members of this cytokine family (namely interleukin 3 and GM-CSF), this glycoprotein in its active form is a homodimer. Interleukin-5 has long been associated with the cause of several allergic diseases including allergic rhinitis and asthma, wherein a large increase in the number of circulating, airway tissue, and induced sputum eosinophils have been observed. Given the high concordance of eosinophils and, in particular, allergic asthma pathology, it has been widely speculated that eosinophils have an important role in the pathology of this disease. Drugs that target IL-5 are mepolizumab and reslizumab.
This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 15.0 kDa. The protein migrates as 18-23 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Especificaciones
NP_000870.1 | |
Human IL-5, His Tag (IL5-H52H3) is expressed from human 293 cells (HEK293). It contains AA Ile 20 - Ser 134 (Accession # P05113-1). | |
-20°C | |
IL-5 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Cytokines and Growth Factors |
15.0 kDa | |
Ile 20 - Ser 134 | |
IL-5,TRF,IL5,Interleukin-5 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |
El Programa Encompass de Fisher Scientific ofrece artículos que no forman parte de nuestra cartera de distribución. Estos productos no suelen tener imágenes ni descripciones detalladas. Sin embargo, nos comprometemos a mejorar su experiencia de compra. Utilice el siguiente formulario para hacernos llegar sus comentarios sobre el contenido de este producto.