Learn More
Abnova™ Human GORASP1 Full-length ORF (NP_114105.1, 1 a.a. - 440 a.a.) Recombinant Protein with GST-tag at N-terminal
Descripción
The Golgi complex plays a key role in the sorting and modification of proteins exported from the endoplasmic reticulum. The protein encoded by this gene is a membrane protein involved in establishing the stacked structure of the Golgi apparatus. It is a caspase-3 substrate, and cleavage of this encoded protein contributes to Golgi fragmentation in apoptosis. This encoded protein can form a complex with the Golgi matrix protein GOLGA2, and this complex binds to the vesicle docking protein p115. Several alternatively spliced transcript variants of this gene have been identified, but their full-length natures have not been determined. [provided by RefSeq]
Especificaciones
Especificaciones
| Número de acceso | NP_114105.1 |
| Para utilizar con (aplicación) | Antibody Production, Protein Array, ELISA, Western Blot |
| Formulación | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
| ID de gen (Entrez) | 64689 |
| Peso molecular | 72.9kDa |
| Nombre | GORASP1 (Human) Recombinant Protein (P01) |
| Método de purificación | Glutathione Sepharose 4 Fast Flow |
| Pruebas de control de calidad | 12.5% SDS-PAGE Stained with Coomassie Blue. |
| Cantidad | 10 μg |
| Inmunógeno | MGLGVSAEQPAGGAEGFHLHGVQENSPAQQAGLEPYFDFIITIGHSRLNKENDTLKALLKANVEKPVKLEVFNMKTMRVREVEVVPSNMWGGQGLLGASVRFCSFRRASEQVWHVLDVEPSSPAALAGLRPYTDYVVGSDQILQESEDFFTLIESHEGKPLKLMVYNSKSDSCREVTVTPNAAWGGEGSLGCGIGYGYLHRIPTQPPSYHKKPPGTPPPSALPLGAPPPDALPPGPTPEDSPSLETGSRQSDYMEALLQAPGSSMEDPLPGPGSPSHSAPDPDGLPHFMETPLQPPPPVQRVMDPGFLDVSGISLLDNSNASVWPSLPSSTELTTTAVSTSGPEDICSSSSSHERGGEATWSGSEFEVSFLDSPGAQAQADHLPQLTLPDSLTSAASPEDGLSAELLEAQAEEEPASTEGLDTGTEAEGLDSQAQISTTE |
| Mostrar más |
Sugerencias de productos
Los clientes que vieron este artículo también vieron
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.