missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human CXCL12 (SDF-1b) Recombinant Protein, PeproTech®
Recombinant Protein
Marca: Gibco™ 300-28B-250UG
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Recombinant Human SDF-1beta is an 8.5 kDa protein containing 72 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
CXCL12 is a stromal cell-derived alpha chemokine member of the intercrine family. This gene product and its receptor CXCR4 can activate lymphocytes and have been implicated in the metastasis of some cancers such as breast cancer. Mutations in this gene are associated with resistance to human immunodeficiency virus type 1 infections. Multiple transcript variants encoding different isoforms have been found for this gene.
Especificaciones
P48061 | |
protein with no preservative | |
8.5 kDa | |
250 μg | |
RUO | |
12-O-tetradecanoylphorbol 13-acetate repressed protein 1; C Cmotif chemokine; C x C motif chemokine; CC motif chemokine; CCmotif chemokine; chemokine (C-X-C motif) ligand 12; chemokine (C-X-C motif) ligand 12 (stromal cell-derived factor 1); CXC; CXC motif chemokine; C-X-C motif chemokine 12; C-X-C motif chemokine ligand 12; CXCL; CXCL12; Intercrine reduced in hepatomas; IRH; PBSF; pre-B cell growth-stimulating factor; pre-B-cell growth-stimulating factor; SCYB12; Sdf1; SDF-1; sdf-1 beta; SDF-1 gamma; SDF1A; SDF-1-alpha(3-67); SDF1B; SDF-1-beta(3-72); Stromal cell-derived factor 1; stromal cell-derived factor-1 gamma; thymic lymphoma cell-stimulating factor; TLSF; Tpar1 | |
Cxcl12 | |
Unconjugated | |
KPVSLSYRCP CRFFESHVAR ANVKHLKILN TPNCALQIVA RLKNNNRQVC IDPKLKWIQE YLEKALNKRF KM | |
E. coli | |
≥ 98% by SDS-PAGE gel and HPLC analyses. |
ELISA, Functional Assay, Western Blot | |
6387 | |
Human CXCL12 (SDF-1 b) | |
E. coli | |
<1 EU/ μg | |
CXCL12 beta | |
Determined by its ability to chemoattract activated human T-lymphocytes using a concentration range of 10-50 ng/ml | |
Recombinant | |
-20°C | |
Lyophilized |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Gibco™ Human CXCL12 (SDF-1b) Recombinant Protein, PeproTech® > 250 μg; Unlabeled
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido