Learn More
Gibco™ Human CCL21 (6Ckine) Recombinant Protein, PeproTech®
Recombinant Protein
Marca:  Gibco™ 300-35A-5UG
77.85 EUR hasta el 2025-06-30
Use el código promocional "24108" para aplicar la oferta.
 Ver más versiones de este producto
Código de producto. 17862123
  • 86.50€ / 5 microgramos
Fomente su progreso
Usar código promocional 24108 al validar el pedido
Ver promociones disponibles
Descripción
Recombinant Human Exodus-2 (CCL21) is a 12.2 kDa protein containing 111 amino acid residues. *Human Exodus-2 (Catalog Number 300-35A) has replaced Human Exodus-2 (Catalog Number 300-35). This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
This gene is one of several CC cytokine genes clustered on the p-arm of chromosome 9. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The CC cytokines are proteins characterized by two adjacent cysteines. Similar to other chemokines the protein encoded by this gene inhibits hemopoiesis and stimulates chemotaxis. This protein is chemotactic in vitro for thymocytes and activated T cells, but not for B cells, macrophages, or neutrophils. The cytokine encoded by this gene may also play a role in mediating homing of lymphocytes to secondary lymphoid organs. It is a high affinity functional ligand for chemokine receptor 7 (CCR7) that is expressed on T and B lymphocytes and a known receptor for another member of the cytokine family (small inducible cytokine A19).
TRUSTED_SUSTAINABILITY
Especificaciones
Mostrar más
Certificados
Promociones
Promociones
Título del producto
Gibco™ Human CCL21 (6Ckine) Recombinant Protein, PeproTech® > 5 μg; Unlabeled