Learn More
Gibco™ Human beta-Defensin 1 (36 aa) Recombinant Protein, PeproTech®
Recombinant Protein
Marca: Gibco™ 300-51-20UG
189.90 EUR hasta el 2025-06-30
Use el código promocional "24108" para aplicar la oferta.
Descripción
Recombinant Human BD-1 is a 3.9 kDa protein containing 36 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
Defensins form a family of microbicidal and cytotoxic peptides made by neutrophils. Members of the defensin family are highly similar in protein sequence. This gene encodes defensin, beta 1, an antimicrobial peptide implicated in the resistance of epithelial surfaces to microbial colonization. This gene maps in close proximity to defensin family member, defensin, alpha 1 and has been implicated in the pathogenesis of cystic fibrosis.
Especificaciones
P60022 | |
protein with no preservative | |
3.9 kDa | |
20 μg | |
RUO | |
AW260221; BD1; BD-1; beta defensin 1; beta-defensin 1; beta-defensin-1; DEFB1; DEFB-1; DEFB101; defensin beta 1; defensin, beta 1; HBD1; rBD-1 | |
DEFB1 | |
Unconjugated | |
DHYNCVSSGG QCLYSACPIF TKIQGTCYRG KAKCCK | |
E. coli | |
≥ 98% by SDS-PAGE gel and HPLC analyses. |
Functional Assay | |
1672 | |
Human beta-Defensin 1 (36 aa) | |
E. coli | |
<1 EU/ μg | |
beta-Defensin 1 | |
Determined by its ability to chemoattract CD34+ dendritic cells using a concentration range of 100.0-1000.0 ng/ml | |
Recombinant | |
-20°C | |
Lyophilized |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.