missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human BCCIP (aa 192-287) Control Fragment Recombinant Protein
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
Descripción
Highest antigen sequence indentity to the following orthologs: Mouse (57%), Rat (57%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-65200 (PA5-65200. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
This gene product was isolated on the basis of its interaction with BRCA2 and p21 proteins. It is an evolutionarily conserved nuclear protein with multiple interacting domains. The N-terminal half shares moderate homology with regions of calmodulin and M-calpain, suggesting that it may also bind calcium. Functional studies indicate that this protein may be an important cofactor for BRCA2 in tumor suppression, and a modulator of CDK2 kinase activity via p21. This protein has also been implicated in the regulation of BRCA2 and RAD51 nuclear focus formation, double-strand break-induced homologous recombination, and cell cycle progression. Multiple transcript variants encoding different isoforms have been described for this gene.

Especificaciones
Especificaciones
Número de acceso | Q9P287 |
Concentración | ≥5.0 mg/mL |
Para utilizar con (aplicación) | Blocking Assay, Control |
Formulación | 1 M urea, PBS with no preservative; pH 7.4 |
ID de gen (Entrez) | 56647 |
Nombre | Human BCCIP (aa 192-287) Control Fragment |
Cantidad | 100 μL |
Estado normativo | RUO |
Alias de gen | 1110013J05Rik; Bccip; BCCIPalpha; BCCIPbeta; BRCA2 and CDKN1A interacting protein; BRCA2 and CDKN1A-interacting protein; BRCA2 and Cip1/p21 interacting protein; cdk inhibitor p21 binding protein; p21- and CDK-associated protein 1; Protein TOK-1; TOK1; TOK-1; TOK-1 alpha; TOK-1 beta |
Nombre común | BCCIP |
Mostrar más |
Sugerencias de productos
Customers who viewed this item also viewed.
Viewing 1-4 of
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Invitrogen™ Human BCCIP (aa 192-287) Control Fragment Recombinant Protein >
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido