Learn More
Abnova™ DLEU1 Recombinant Protein
Marca:  Abnova™ H00010301-P01.10ug
 Ver más versiones de este producto
Código de producto. 16164862
  • 344.00€ / 10 microgramos
Descripción
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Especificaciones
Especificaciones
AAH20692
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
34.32
8
Glutathione Sepharose 4 Fast Flow
10 μg
MRPCIWIHVHLKPPCRLVELLPFSSALQGLSHLSLGTTLPVILPERNEEQNLQELSHNADKYQMGDCCKEEIDDSIFY
BCMS/DLB1/DLEU2/LEU1/LEU2/MGC22430/NCRNA00021/XTP6
DLEU1
Wheat Germ (in vitro)
GST
Antibody Production, Protein Array, ELISA, Western Blot
10301
DLEU1 (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DLEU1
Human
Recombinant
Solution
Certificados
Promociones
Promociones
Título del producto
Abnova™ DLEU1 Recombinant Protein > 10μg