Learn More
Cediranib, MedChemExpress
MedChemExpress Cediranib (AZD2171) is a highly potent, orally available VEGFR tyrosine kinase inhibitor with IC50s of <1, <3, 5, 5, 36, 2 nM for Flt1, KDR, Flt4, PDGFRα, PDGFRβ, c-Kit, respectively.
Marca: Medchem Express HY-10205
Descripción
Cediranib (AZD2171) is a highly potent, orally available VEGFR tyrosine kinase inhibitor with IC50s of <1, <3, 5, 5, 36, 2 nM for Flt1, KDR, Flt4, PDGFRα, PDGFRβ, c-Kit, respectively.Especificaciones
Cediranib | |
Off-White | |
C25H27FN4O3 | |
AZD2171 | |
FC1=C(OC2=C(C(C=C3OCCCN4CCCC4)=NC=N2)C=C3OC)C=CC5=C1C=C(C)N5 | |
450.51 | |
Research | |
Cancer-Kinase/protease |
288383-20-0 | |
Research | |
50 mg | |
DMSO : 20 mg/mL (44.39 mM; Need ultrasonic) | |
450.51 | |
98.0% | |
Solid |