Learn More
TdB Labs AB 500 kDa Blue Dextran for SEC-chromatography and Macromolecular Tracing
Blue dextran 500 kDa is designed for studying the upper limits of vascular or gel permeability. It serves as a high MW tracer in biomedical diffusion studies and chromatography calibration.

125.90€ - 679.59€
Especificaciones
Color | Blue |
---|---|
Formato | Ready-to-use |
Tipo de producto | Protein Molecular Weight Marker |
Intervalo de tamaños | 40 to 500 kDa |
Peso molecular | 500 kDa |
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
30216274
![]() |
TdB Labs AB
BD500_1g |
1 g |
125.90€
1 g |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | |||||
30216506
![]() |
TdB Labs AB
BD500_10g |
10 g |
679.59€
10 g |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | |||||
Descripción
- Product Overview:Blue dextran is a non-fluorescent, water-soluble polysaccharide available in molecular weights ranging from 5 kDa to 2000 kDa. It is commonly used in permeability assays, chromatography, and fluid dynamics studies, offering flexibility for a range of experimental applications.
- Features:
- Non-fluorescent, stable polymer, ideal for macromolecular transport and barrier permeability studies.
- Available in molecular weights from 5 kDa to 2000 kDa, providing flexibility to suit different experimental needs.
- Water-soluble, making it easy to incorporate into aqueous biological systems for transport and volume studies.
- High stability and long shelf life, ensuring consistent results over extended periods.
- Benefits:
- Versatile Molecular Weight Range:Blue dextran's availability in multiple molecular weights (5, 10, 20, 40, 70, 110, 500, and 2000 kDa) allows for precise experimental design, from small molecular studies to large molecule transport.
- Stable and Non-toxic: Safe for use in both in vitro and in vivo models without affecting biological systems, making it ideal for permeability and volume studies.
- Reliable and Reproducible:Blue dextran offers consistent performance over time, with long-term stability in storage, ensuring reproducibility in experimental settings.
- Applications:
- Used in permeability assays to assess the passage of macromolecules across biological barriers, including cell monolayers, tissues, and capillaries.
- Commonly applied in chromatography for column calibration and determination of molecular weight exclusion limits.
- Used in blood volume studies to measure plasma volume in clinical and preclinical research.
- Effective in vascular studies to track macromolecule retention and flow through microvascular networks.
- Technical Specifications:
- Molecular Weights: Available in 5 kDa, 10 kDa, 20 kDa, 40 kDa, 70 kDa, 110 kDa, 500 kDa, and 2000 kDa to meet diverse experimental requirements.
- Solubility: Fully soluble in water, facilitating easy integration into aqueous-based experimental systems.
- Storage and Stability:
- Stable for extended periods when stored dry in sealed containers at ambient temperature.
- Protect from moisture and light to maintain optimal stability and effectiveness.
- Packaging and Delivery:
- Blue Dextran is supplied as a powder, ensuring long shelf life and easy handling.
- Available in various packaging sizes, ranging from small quantities for initial studies to bulk packaging for large-scale applications.
Especificaciones
Blue | |
Protein Molecular Weight Marker | |
500 kDa |
Ready-to-use | |
40 to 500 kDa |
El Programa Encompass de Fisher Scientific ofrece artículos que no forman parte de nuestra cartera de distribución. Estos productos no suelen tener imágenes ni descripciones detalladas. Sin embargo, nos comprometemos a mejorar su experiencia de compra. Utilice el siguiente formulario para hacernos llegar sus comentarios sobre el contenido de este producto.