missing translation for 'onlineSavingsMsg'
Learn More

TdB Labs AB 500 kDa Blue Dextran for SEC-chromatography and Macromolecular Tracing

Código de producto. 30216274
Encompass_Preferred
Change view
Click to view available options
Cantidad:
1 g
10 g
Tamaño de la unidad:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
30216274 1 g 1 g
30216506 10 g 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 30216274 Proveedor TdB Labs AB N.º de proveedor BD500_1g

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Blue dextran 500 kDa is designed for studying the upper limits of vascular or gel permeability. It serves as a high MW tracer in biomedical diffusion studies and chromatography calibration.

  • Product Overview:Blue dextran is a non-fluorescent, water-soluble polysaccharide available in molecular weights ranging from 5 kDa to 2000 kDa. It is commonly used in permeability assays, chromatography, and fluid dynamics studies, offering flexibility for a range of experimental applications.
  • Features:
    • Non-fluorescent, stable polymer, ideal for macromolecular transport and barrier permeability studies.
    • Available in molecular weights from 5 kDa to 2000 kDa, providing flexibility to suit different experimental needs.
    • Water-soluble, making it easy to incorporate into aqueous biological systems for transport and volume studies.
    • High stability and long shelf life, ensuring consistent results over extended periods.
  • Benefits:
    • Versatile Molecular Weight Range:Blue dextran's availability in multiple molecular weights (5, 10, 20, 40, 70, 110, 500, and 2000 kDa) allows for precise experimental design, from small molecular studies to large molecule transport.
    • Stable and Non-toxic: Safe for use in both in vitro and in vivo models without affecting biological systems, making it ideal for permeability and volume studies.
    • Reliable and Reproducible:Blue dextran offers consistent performance over time, with long-term stability in storage, ensuring reproducibility in experimental settings.
  • Applications:
    • Used in permeability assays to assess the passage of macromolecules across biological barriers, including cell monolayers, tissues, and capillaries.
    • Commonly applied in chromatography for column calibration and determination of molecular weight exclusion limits.
    • Used in blood volume studies to measure plasma volume in clinical and preclinical research.
    • Effective in vascular studies to track macromolecule retention and flow through microvascular networks.
  • Technical Specifications:
    • Molecular Weights: Available in 5 kDa, 10 kDa, 20 kDa, 40 kDa, 70 kDa, 110 kDa, 500 kDa, and 2000 kDa to meet diverse experimental requirements.
    • Solubility: Fully soluble in water, facilitating easy integration into aqueous-based experimental systems.
  • Storage and Stability:
    • Stable for extended periods when stored dry in sealed containers at ambient temperature.
    • Protect from moisture and light to maintain optimal stability and effectiveness.
  • Packaging and Delivery:
    • Blue Dextran is supplied as a powder, ensuring long shelf life and easy handling.
    • Available in various packaging sizes, ranging from small quantities for initial studies to bulk packaging for large-scale applications.

Especificaciones

Color Blue
Formato Ready-to-use
Intervalo de tamaños 40 to 500 kDa
Peso molecular 500 kDa
Cantidad 1 g
Tipo de producto Protein Molecular Weight Marker
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.