missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Paragon Scientific™ Viscosity Bath Media; White Oil for use 40°C to 80°C
Formulated using high quality raw materials to facilitate optimum visibility, maximum heat stability and efficiency of thermostatic baths. Low viscosity aids dissipation of air bubbles which can lead to detection problems in automatic systems and ensures bath stability and uniformity are optimized.
76.85€ - 200.00€
Código de producto | Marca | Capacidad (métrico) | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Capacidad (métrico) | Precio | Cantidad y disponibilidad | |||||
16000172
|
Paragon Scientific™
BM1-5L |
5 l |
76.85€
5 litros |
Inicie sesión para ver el stock |
16090162
|
Paragon Scientific™
BM1-20L |
20 l |
200.00€
20 litros |
Inicie sesión para ver el stock |
Descripción
- Paragon Scientific Bath Media
- Viscosity Bath Media; White Oil for use 40°C to 80°C
- Tamper evident security packaging
- Manufactured in the United kingdom
Especificaciones
De 40 °C a 80 °C |