Learn More
TRA-1-60 (Podocalyxin) Mouse, anti-Human, PE, Clone: TRA-1-60R, STEMCELL Technologies
Mouse Monoclonal Antibody
Marca: Stemcell Technologies 60064PE.1
Descripción
- The TRA‐1‐60R antibody reacts with TRA-1-60 (Podocalyxin), a > 200 kDa pluripotent stem cell-specific protein expressed on the surface of undifferentiated human embryonic stem (ES) and induced pluripotent stem (iPS) cells, embryonal carcinoma (EC) cells and embryonic germ (EG) cells, as well as rhesus monkey ES cell lines
- The epitope contains sialic acid and is associated with a large-molecular-mass transmembrane protein named podocalyxin
- Though sialylated, the epitope recognized by the TRA‐1‐60R antibody is resistant to treatment with neuraminidase
- This antibody clone has been verified for labeling human ES and iPS cells grown in TeSR™-E8™ (Catalog #05940), mTeSR™1 (Catalog #85850), and TeSR™2 (Catalog #05860)
Especificaciones
TRA-1-60 (Podocalyxin) | |
Monoclonal | |
PE | |
25 Tests | |
Rhesus Monkey, Human, Rabbit | |
Pluripotent Stem Cells |
Immunoprecipitation, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
TRA-1-60R | |
Mouse | |
Primary | |
IgM κ |
El Programa Encompass de Fisher Scientific ofrece artículos que no forman parte de nuestra cartera de distribución. Estos productos no suelen tener imágenes ni descripciones detalladas. Sin embargo, nos comprometemos a mejorar su experiencia de compra. Utilice el siguiente formulario para hacernos llegar sus comentarios sobre el contenido de este producto.