Learn More
TLR7/8 agonist 1 dihydrochloride, MedChemExpress
MedChemExpress TLR7/8 agonist 1 dihydrochloride is a toll-like receptor TLR7/TLR8 dual-agonistic imidazoquinoline.
Marca: Medchem Express HY-103698A
Descripción
TLR7/8 agonist 1 dihydrochloride is a toll-like receptor TLR7/TLR8 dual-agonistic imidazoquinoline.Especificaciones
TLR7/8 agonist 1 dihydrochloride | |
Light Yellow | |
C22H27Cl2N5 | |
DMSO : 83.33 mg/mL (192.72 mM; Need ultrasonic) | |
432.39 | |
98.0% | |
Solid |
1620278-72-9 | |
Research | |
100 mg | |
NC1=NC2=CC=CC=C2C3=C1N=C(CCCC)N3CC4=CC=C(CN)C=C4.[H]Cl.[H]Cl | |
432.39 | |
Research | |
COVID-19-immunoregulation |