Learn More
Terlipressin, MedChemExpress
MedChemExpress Terlipressin is a vasopressin analogue with potent vasoactive properties. Terlipressin is a highly selective vasopressin V1 receptor agonist that reduces the splanchnic blood flow and portal pressure and controls acute variceal bleeding. Terlipressin exerts anti-inflammatory and anti-oxidative effects. Terlipressin has the potential for hepatorenal syndrome and norepinephrine-resistant septic shock research.
Marca: Medchem Express HY-12554
Descripción
Terlipressin is a vasopressin analogue with potent vasoactive properties. Terlipressin is a highly selective vasopressin V1 receptor agonist that reduces the splanchnic blood flow and portal pressure and controls acute variceal bleeding. Terlipressin exerts anti-inflammatory and anti-oxidative effects. Terlipressin has the potential for hepatorenal syndrome and norepinephrine-resistant septic shock research.Especificaciones
Terlipressin | |
Research | |
5 mg | |
O=C([C@H](CSSC[C@@H](C(N[C@H](C1=O)CC2=CC=C(O)C=C2)=O)NC(CNC(CNC(CN)=O)=O)=O)NC([C@@H](NC([C@@H](NC([C@](N1)([H])CC3=CC=CC=C3)=O)CCC(N)=O)=O)CC(N)=O)=O)N(CCC4)[C@@H]4C(N[C@@H](CCCCN)C(NCC(N)=O)=O)=O | |
1227.37 | |
COVID-19-immunoregulation |
14636-12-5 | |
C52H74N16O15S2 | |
DMSO : 100 mg/mL (81.48 mM; Need ultrasonic) | |
1227.37 | |
Research |