Learn More
Notrax Sof-Tred™ Microcellular Vinyl Foam with Pebble Pattern, Gray
Notrax Sof-Tred™ Basic worker comfort derived from 9.4 mm thick worker platform comprised of a construction of microcellular vinyl consisting of millions of air bubbles that provide basic ergonomic benefit.
Marca:  Notrax 411C0348GY
Detalles adicionales : Peso : 2.80000kg
 Ver más versiones de este producto
Código de producto. 17792078
  • 62.12€ / cada uno
Fecha estimada de envío: 18-04-2023
para ver el stock
Añadir a la cesta
Descripción
Descripción
  • Pebble pattern provides non-directional traction and is easy to clean
  • Basic worker comfort derived from 94 mm thick worker platform comprised of a construction of microcellular vinyl consisting of millions of air bubbles that provide basic ergonomic benefit
  • All four sides are bevelled and sealed to minimize tripping hazards
Especificaciones
Especificaciones
122 cm
Microcellular Vinyl
60 ft.
Industrial and Commercial Matting
1830 cm
Gray