missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SETD7 Monoclonal Antibody (7C1B7E1), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA543500
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Specificity: Human SETD7/SET7/9
Histone methyltransferase that specifically monomethylates 'Lys-4' of histone H3. H3 'Lys-4' methylation represents a specific tag for epigenetic transcriptional activation. Plays a central role in the transcriptional activation of genes such as collagenase or insulin. Recruited by IPF1/PDX-1 to the insulin promoter, leading to activate transcription. Has also methyltransferase activity toward non-histone proteins such as p53/TP53, TAF10, and possibly TAF7 by recognizing and binding the [KR]-[STA]-K in substrate proteins. Monomethylates 'Lys-189' of TAF10, leading to increase the affinity of TAF10 for RNA polymerase II. Monomethylates 'Lys-372' of p53/TP53, stabilizing p53/TP53 and increasing p53/TP53-mediated transcriptional activation. [UniProt]
Especificaciones
SETD7 | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
Q8WTS6 | |
Setd7 | |
Recombinant Human SETD7 protein. | |
50 μL | |
Primary | |
Human | |
Antibody | |
IgG2b |
ELISA | |
7C1B7E1 | |
Unconjugated | |
Setd7 | |
1600028F23Rik; FLJ21193; H3-K4 methyltransferase; H3-K4-HMTase; H3-K4-HMTase SETD7; H3K4MT; Histone H3-K4 methyltransferase SETD7; histone H3-lysine 4-specific methyltransferase; histone-lysine N-methyltransferase SETD7; Kiaa1717; KMT7; lysine N-methyltransferase 7; mKIAA1717; OTTHUMP00000220049; SET domain containing (lysine methyltransferase) 7; SET domain containing 7, histone lysine methyltransferase; SET domain containing 7, lysine methyltransferase; SET domain containing lysine methyltransferase 7; SET domain-containing protein 7; Set7; Set7/9; SET9; Setd7 | |
Mouse | |
Protein A | |
RUO | |
80854 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
SETD7 Monoclonal Antibody (7C1B7E1), Invitrogen™ > 50 μL; Unconjugated
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido